Thermo Scientific Chemicals

1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals

Catalog number: A11395.30
250 g, Each
Thermo Scientific Chemicals

1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals

Catalog number: A11395.30
250 g, Each
Quantity
Catalog number: A11395.30
also known as A11395-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-70-6
IUPAC Name
1-bromo-3-chloropropane
Molecular Formula
C3H6BrCl
InChI Key
MFESCIUQSIBMSM-UHFFFAOYSA-N
SMILES
ClCCCBr
Form
Liquid
Comment
Material sourced in UK and US
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5% (UK-sourced material)
Refractive Index
1.4845-1.4875 @ 20?C (UK-sourced material)

Description

1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.

Solubility
Miscible with alcohols, chlorofom and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text