2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals
2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A11403.14
also known as A11403-14
Price (USD)/ Each
201.65
Online exclusive
224.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
201.65
Online exclusive
224.00 
Save 22.35 (10%)
Add to cart
2,2,2-Trifluoroacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA11403.14
Price (USD)/ Each
201.65
Online exclusive
224.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS434-45-7
IUPAC Name2,2,2-trifluoro-1-phenylethan-1-one
Molecular FormulaC8H5F3O
InChI KeyKZJRKRQSDZGHEC-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4560-1.4600 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only