Thermo Scientific Chemicals

Hydroquinone, 99%, Thermo Scientific Chemicals

Catalog number: A11411.30
250 g, Each
Thermo Scientific Chemicals

Hydroquinone, 99%, Thermo Scientific Chemicals

Catalog number: A11411.30
250 g, Each
Quantity
Catalog number: A11411.30
also known as A11411-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-31-9
IUPAC Name
benzene-1,4-diol
Molecular Formula
C6H6O2
InChI Key
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILES
OC1=CC=C(O)C=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
170.0-176.0?C
Identification (FTIR)
Conforms

Description

Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.

Solubility
Soluble in water, methanol, ethanol, diethyl ether, acetone, benzene and carbon tetrachloride.

Notes
Air and light sensitive. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text