1,1,3,3-Tetramethoxypropane, 98%, Thermo Scientific Chemicals
1,1,3,3-Tetramethoxypropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,3,3-Tetramethoxypropane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A11418.30
also known as A11418-30
Price (USD)/ Each
44.65
Online exclusive
49.90 
Save 5.25 (11%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.90 
Save 5.25 (11%)
Add to cart
1,1,3,3-Tetramethoxypropane, 98%, Thermo Scientific Chemicals
Catalog numberA11418.30
Price (USD)/ Each
44.65
Online exclusive
49.90 
Save 5.25 (11%)
-
Add to cart
Chemical Identifiers
CAS102-52-3
IUPAC Name1,1,3,3-tetramethoxypropane
Molecular FormulaC7H16O4
InChI KeyXHTYQFMRBQUCPX-UHFFFAOYSA-N
SMILESCOC(CC(OC)OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4050-1.4090 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only