Thermo Scientific Chemicals

Benzopinacol, 98%, Thermo Scientific Chemicals

Catalog number: A11421.14
25 g, Each
Thermo Scientific Chemicals

Benzopinacol, 98%, Thermo Scientific Chemicals

Catalog number: A11421.14
25 g, Each
Quantity
Catalog number: A11421.14
also known as A11421-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
464-72-2
IUPAC Name
tetraphenylethane-1,2-diol
Molecular Formula
C26H22O2
InChI Key
MFEWNFVBWPABCX-UHFFFAOYSA-N
SMILES
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to very pale cream or very pale grey
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications: Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text