Thermo Scientific Chemicals

1,3-Phenylenediacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11429.14
25 g, Each
Thermo Scientific Chemicals

1,3-Phenylenediacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11429.14
25 g, Each
Quantity
Catalog number: A11429.14
also known as A11429-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19806-17-8
Form
Crystals and/or powder
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%
Assay (HPLC)
≥96.0%
Appearance (Color)
White to cream
Melting Point (clear melt)
171-179?C

Description

In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1?-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1′-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

Solubility
Slightly soluble in water. Solublitiy in methanol is almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text