Thermo Scientific Chemicals

5-Aminonaphthalene-1-sulfonic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A11433.14
25 g, Each
Thermo Scientific Chemicals

5-Aminonaphthalene-1-sulfonic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A11433.14
25 g, Each
Quantity
Catalog number: A11433.14
also known as A11433-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84-89-9
IUPAC Name
5-aminonaphthalene-1-sulfonic acid
Molecular Formula
C10H9NO3S
InChI Key
DQNAQOYOSRJXFZ-UHFFFAOYSA-N
SMILES
NC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O
Assay (Aqueous acid-base Titration)
≥80.0%
Appearance (Color)
Pale purple to grey or brown to dark brown
Form
Powder

Description

5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminonaphthalene-1-sulfonic acid is an deriva- tizing agents in the precolumn derivatization of some chiral phenoxy acid. Cu(II) based on a dansyl derivative was synthesized with 29.2% overall yield by a seven-step synthetic procedure from the readily available starting material 5-aminonaphthalene-1-sulfonic acid.

Solubility
Soluble in hot water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text