2,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
2,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1000 g
Catalog number A11435.36
also known as A11435-36
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
2,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA11435.36
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS94-99-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5740-1.5780 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only