Thermo Scientific Chemicals

2-(2-Bromoethyl)-1,3-dioxolane, 95%, stab. with silver, Thermo Scientific Chemicals

Catalog number: A11441.18
50 g, Each
Thermo Scientific Chemicals

2-(2-Bromoethyl)-1,3-dioxolane, 95%, stab. with silver, Thermo Scientific Chemicals

Catalog number: A11441.18
50 g, Each
Quantity
Catalog number: A11441.18
also known as A11441-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18742-02-4
IUPAC Name
2-(2-bromoethyl)-1,3-dioxolane
Molecular Formula
C5H9BrO2
InChI Key
GGZQLTVZPOGLCC-UHFFFAOYSA-N
SMILES
BrCCC1OCCO1
Assay (GC)
≥94.0%
Refractive Index
1.4760-1.4820 @ 20?C
Appearance (Color)
Clear colorless to yellow or pale brown
Identification (FTIR)
Conforms
Form
Liquid

Description

2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromoethyl)-1,3-dioxolane is used as a pharmaceutical intermediate, dioxolanes, acetals, ketals, building blocks, chemical synthesis, organic building blocks and oxygen compounds. 1,1-(Ethylenedioxy)-3-bromopropane is used in the synthesis of EGFR inhibitors. Also used in the synthesis of orally active agents of cancer and cell proliferation with quinoline substructures. It is also used as flavouring agent.

Solubility
It is immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text