Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
Solubility
It is insoluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Bjorn Akermark.; Lennart Eberson.; Ernst Jonsson.; Egon Pettersson. Palladium-promoted cyclization of diphenyl ether, diphenylamine, and related compounds. J. Org. Chem.197540, (9), 1365-1367.
Frederick G. Bordwell.; George E. Drucker.; Herbert E. Fried. Acidities of carbon and nitrogen acids: the aromaticity of the cyclopentadienyl anion. J. Org. Chem.198146, (3), 632-635.