Thermo Scientific Chemicals

Carbazole, 95%, Thermo Scientific Chemicals

Catalog number: A11448.36
500 g, Each
Thermo Scientific Chemicals

Carbazole, 95%, Thermo Scientific Chemicals

Catalog number: A11448.36
500 g, Each
Quantity
Catalog number: A11448.36
also known as A11448-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-74-8
IUPAC Name
9H-carbazole
Molecular Formula
C12H9N
InChI Key
UJOBWOGCFQCDNV-UHFFFAOYSA-N
SMILES
N1C2=C(C=CC=C2)C2=C1C=CC=C2
Melting Point (clear melt)
238.0-250.0?C
Assay (GC)
≥94.0%
Appearance (Color)
White to cream to yellow to brown to pale gray
Form
Crystals or powder or crystalline powder or flakes or chunks
Identification (FTIR)
Conforms

Description

Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

Solubility
It is insoluble in water.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text