Thermo Scientific Chemicals

Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals

Catalog number: A11450.22
100 g, Each
Thermo Scientific Chemicals

Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals

Catalog number: A11450.22
100 g, Each
Quantity
Catalog number: A11450.22
also known as A11450-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-89-6
IUPAC Name
potassium (ethoxymethanethioyl)sulfanide
Molecular Formula
C3H5KOS2
InChI Key
JCBJVAJGLKENNC-UHFFFAOYSA-M
SMILES
[K+].CCOC([S-])=S
Appearance (Color)
Pale cream to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (Iodometric Titration)
≥97.5 to ≤102.5% (Dry wt. basis)
Identification (FTIR)
Conforms
Loss on Drying
≤2% (105°C/constant weight)

Description

Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text