Thermo Scientific Chemicals

DL-Methionine, 99%

Catalog number: A11457.0B
1000 g, Each
Thermo Scientific Chemicals

DL-Methionine, 99%

Catalog number: A11457.0B
1000 g, Each
Quantity
Catalog number: A11457.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-51-8
IUPAC Name
2-amino-4-(methylsulfanyl)butanoic acid
Molecular Formula
C5H11NO2S
InChI Key
FFEARJCKVFRZRR-UHFFFAOYNA-N
SMILES
CSCCC(N)C(O)=O
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text