Thermo Scientific Chemicals

Dibenzyl carbonate, 98%, Thermo Scientific Chemicals

Catalog number: A11464.30
250 g, Each
Thermo Scientific Chemicals

Dibenzyl carbonate, 98%, Thermo Scientific Chemicals

Catalog number: A11464.30
250 g, Each
Quantity
Catalog number: A11464.30
also known as A11464-30
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
3459-92-5
IUPAC Name
dibenzyl carbonate
Molecular Formula
C15H14O3
InChI Key
PIZLBWGMERQCOC-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to white to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5460-1.5500 @ 20?C

Description

Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

Solubility
Sparingly Soluble (0.099 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 - Harmful if swallowed or in contact with skin

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water