3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11474.14
also known as A11474-14
Price (USD)/ Each
150.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
150.00
Add to cart
3-Amino-2-cyclohexen-1-one, 98% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Catalog numberA11474.14
Price (USD)/ Each
150.00
-
Add to cart
Chemical Identifiers
CAS5220-49-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale brown
FormCrystalline powder and/or chunks
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)<5.0%
Melting Point120-134?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only