Thermo Scientific Chemicals

3-Iodo-4-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11485.14
25 g, Each
Thermo Scientific Chemicals

3-Iodo-4-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11485.14
25 g, Each
Quantity
Catalog number: A11485.14
also known as A11485-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35944-64-0
Appearance (Color)
Brown to dark brown
Form
Crystals or crystalline powder or fused solid
Assay (GC)
≥97.5%
Melting Point (clear melt)
36.0-42.0°C

Description

3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylaniline is used as fine chemicals, pharmaceutical intermediates. 2-[((3-iodo-4-methyl)phenylimino)methyl]-5-nitrothiophene was derived from 5-nitro-2-thiophene-carboxaldehyde and 3-iodo-4-methylaniline. 1-(3-iodo-4-methylphenyl)-3-tert-butyl-2H-aminopyrazole (5), prepared from 3-iodo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Light Sensitive .Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text