Thermo Scientific Chemicals

alpha-Chloralose, 98+%, beta anomer ca 20%, Thermo Scientific Chemicals

Catalog number: A11492.22
100 g, Each
Thermo Scientific Chemicals

alpha-Chloralose, 98+%, beta anomer ca 20%, Thermo Scientific Chemicals

Catalog number: A11492.22
100 g, Each
Quantity
Catalog number: A11492.22
also known as A11492-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15879-93-3
Assay (GC)
≤20% (Beta chloralose)
Optical Rotation
+15? to +19? (c= 2 in Ethanol)
Appearance (Color)
White to pale yellow
Melting Point (clear melt)
174-185?C
Form
Powder

Description

It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canadensis) and blue geese (Chen caerulescens) were caught with alpha-chloralose, an oral hypnotic administered on bait. Wild turkeys (Meleagris gallopavo) were captured with alpha-chloralose, an oral anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed widely as an animal anesthetic in the laboratory setting. alpha-chloralose is a true anesthetic or an immobilizing agent with sedative-hypnotic properties has important implications. Canada geese (Branta canaden
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text