Thermo Scientific Chemicals

1-Chlorobutane, 99+%, Thermo Scientific Chemicals

Catalog number: A11494.0F
2500 mL, Each
Thermo Scientific Chemicals

1-Chlorobutane, 99+%, Thermo Scientific Chemicals

Catalog number: A11494.0F
2500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11494.0F
also known as A11494-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-69-3
IUPAC Name
1-chlorobutane
Molecular Formula
C4H9Cl
InChI Key
VFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILES
CCCCCl
Appearance (Color)
Clear colorless
Refractive Index
1.4010-1.4035 @ 20?C
Assay (GC)
≥99.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text