Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: A11498.36
500 g, Each
Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: A11498.36
500 g, Each
Quantity
Catalog number: A11498.36
also known as A11498-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-56-6
IUPAC Name
ethyl 2-cyanoacetate
Molecular Formula
C5H7NO2
InChI Key
ZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILES
CCOC(=O)CC#N
Form
Liquid
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4160-1.4200 @ 20?C

Description

Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

Solubility
It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform, ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials strong acids, strong bases, strong oxidizing agents, strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text