Thermo Scientific Chemicals

p-Tolyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11499.06
5 g, Each
Thermo Scientific Chemicals

p-Tolyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11499.06
5 g, Each
Quantity
Catalog number: A11499.06
also known as A11499-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-59-3
IUPAC Name
1-isothiocyanato-4-methylbenzene
Molecular Formula
C8H7NS
InChI Key
ABQKHKWXTUVKGF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)N=C=S
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.6330-1.6380 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

Solubility
Decompose in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text