Thermo Scientific Chemicals

Hexafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11500.30
250 g, Each
Thermo Scientific Chemicals

Hexafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11500.30
250 g, Each
Quantity
Catalog number: A11500.30
also known as A11500-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
392-56-3
IUPAC Name
hexafluorobenzene
Molecular Formula
C6F6
InChI Key
ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(F)C(F)=C1F
Refractive Index
1.3760-1.3790 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid
Identification (FTIR)
Conforms

Description

Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. Further, it is used as a reference molecule to investigate tissue oxygenation studies. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text