Thermo Scientific Chemicals

4-Bromocinnamic acid, predominantly trans, 97+%, Thermo Scientific Chemicals

Catalog number: A11503.06
5 g, Each
Thermo Scientific Chemicals

4-Bromocinnamic acid, predominantly trans, 97+%, Thermo Scientific Chemicals

Catalog number: A11503.06
5 g, Each
Quantity
Catalog number: A11503.06
also known as A11503-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1200-07-3
IUPAC Name
(2E)-3-(4-bromophenyl)prop-2-enoic acid
Molecular Formula
C9H7BrO2
InChI Key
CPDDDTNAMBSPRN-ZZXKWVIFSA-N
SMILES
OC(=O)C=CC1=CC=C(Br)C=C1
Melting Point (clear melt)
259.5-265.5°C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.0%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%

Description

4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.

Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text