Thermo Scientific Chemicals

p-Toluic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11506.0I
5000 g, Each
Thermo Scientific Chemicals

p-Toluic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11506.0I
5000 g, Each
Quantity
Catalog number: A11506.0I
also known as A11506-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-94-5
IUPAC Name
4-methylbenzoic acid
Molecular Formula
C8H8O2
InChI Key
LPNBBFKOUUSUDB-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
176.0-184.0?C
Form
Crystals or powder or crystalline powder or flakes
Assay (Silylated GC)
≥97.5%
Appearance (Color)
White to cream

Description

It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in some industrial processes for the oxidation of p-xylene to terephthalic acid which is used in the manufacture of polyethylene terephthalate.  It is also used in medicine, agricultural chemicals, API, dye intermediate. p-Toluic Acid is used as an intermediate for polymer stabilizers, light sensitive compounds, animal feed supplements and other organic chemicals (pharmaceuticals, pigments and dyestuff). They are also used as intermediates for the manufacture of a variety of target compounds.

Solubility
It is insoluble in water, but soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text