3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A11507.06
also known as A11507-06
Price (USD)/ Each
197.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
197.00
Add to cart
3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberA11507.06
Price (USD)/ Each
197.00
-
Add to cart
Chemical Identifiers
CAS2905-24-0
IUPAC Name3-bromobenzene-1-sulfonyl chloride
Molecular FormulaC6H4BrClO2S
InChI KeyPJGOLCXVWIYXRQ-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5895-1.5945 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only