3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in Chloroform and Methanol. Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.
RUO – Research Use Only
M Pumera; I Jelínek; J Jindrich. Determination of cyclodextrins and their derivatives by capillary electrophoresis with indirect UV and conductivity detection. Fresenius’ Journal of Analytical Chemistry.2001, 369 (7-8), 666-669.
G Vaidyanathan; M R Zalutsky. Radioiodination of antibodies via N-succinimidyl 2,4-dimethoxy-3-(trialkylstannyl)benzoates. Bioconjugate Chemistry.1990, 1 (6), 387-393.