Thermo Scientific Chemicals

3-Iodobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11515.18
50 g, Each
Thermo Scientific Chemicals

3-Iodobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11515.18
50 g, Each
Quantity
Catalog number: A11515.18
also known as A11515-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-51-9
IUPAC Name
3-iodobenzoic acid
Molecular Formula
C7H5IO2
InChI Key
KVBWBCRPWVKFQT-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(I)=C1
Appearance (Color)
White to cream to pale yellow or pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%

Description

3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in Chloroform and Methanol. Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text