Thermo Scientific Chemicals

1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals

Catalog number: A11516.03
1 g, Each
Thermo Scientific Chemicals

1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals

Catalog number: A11516.03
1 g, Each
Quantity
Catalog number: A11516.03
also known as A11516-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
295-37-4
IUPAC Name
1,4,8,11-tetraazacyclotetradecane
Molecular Formula
C10H24N4
InChI Key
MDAXKAUIABOHTD-UHFFFAOYSA-N
SMILES
C1CNCCNCCCNCCNC1
Form
Fibrous crystalline solid or crystals or needles or powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
183-192?C
Appearance (Color)
White to pale cream
Water Content (Karl Fischer Titration)
<2.0%

Description

1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text