Thermo Scientific Chemicals

Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals

Catalog number: A11520.30
250 g, Each
Thermo Scientific Chemicals

Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals

Catalog number: A11520.30
250 g, Each
Quantity
Catalog number: A11520.30
also known as A11520-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
383-63-1
IUPAC Name
ethyl 2,2,2-trifluoroacetate
Molecular Formula
C4H5F3O2
InChI Key
STSCVKRWJPWALQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C(F)(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.3060-1.3090 @ 20?C
Identification (FTIR)
Conforms

Description

Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

Solubility
Slightly miscible with water. Miscible with chloroform and methanol.

Notes
Moisture Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text