Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
RUO – Research Use Only
Kyrre Thalberg.; Bjoern Lindman. Interaction between hyaluronan and cationic surfactants. J. Phys. Chem. 1989, 93, (4), 1478-1483.
Herbert O. House.; Edith Feng.; Norton P. Peet. Comparison of various tetraalkylammonium salts as supporting electrolytes in organic electrochemical reactions. J. Org. Chem. 1971, 36, (16), 2371-2375.