Thermo Scientific Chemicals

Malonic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11526.22
100 g, Each
Thermo Scientific Chemicals

Malonic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11526.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11526.22
also known as A11526-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-82-2
IUPAC Name
propanedioic acid
Molecular Formula
C3H4O4
InChI Key
OFOBLEOULBTSOW-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or granules
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries

Description

Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

Solubility
Soluble in cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text