Thermo Scientific Chemicals

Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11528.22
100 g, Each
Thermo Scientific Chemicals

Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11528.22
100 g, Each
Quantity
Catalog number: A11528.22
also known as A11528-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-73-6
IUPAC Name
N,2-dihydroxybenzamide
Molecular Formula
C7H7NO3
InChI Key
HBROZNQEVUILML-UHFFFAOYSA-N
SMILES
ONC(=O)C1=CC=CC=C1O
Appearance (Color)
Pale pink to pink
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (HPLC)
≥98.5%

Description

It is widely used for a variety of roles in biology and medicine as a chelating therapy. It inhibits bacterial or fungi growth by interfering with iron uptake. It is also active as a inhibitor of enzyme involved in tumour growths. These compounds are used as extractants for the separation of valuable metals or minerals including copper, lead, gold, other rare earth metals and kaolin. Hydroxamic acids are also used to produce corresponding isocyanates by dehydration followed by rearrangement reaction. It is also used as a common ligand utilized in the synthesis of metallacrowns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used for a variety of roles in biology and medicine as a chelating therapy. It inhibits bacterial or fungi growth by interfering with iron uptake. It is also active as a inhibitor of enzyme involved in tumour growths. These compounds are used as extractants for the separation of valuable metals or minerals including copper, lead, gold, other rare earth metals and kaolin. Hydroxamic acids are also used to produce corresponding isocyanates by dehydration followed by rearrangement reaction. It is also used as a common ligand utilized in the synthesis of metallacrowns.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong bases. Stable under normal conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text