Thermo Scientific Chemicals

Tetraphenylphosphonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A11539.22
100 g, Each
Thermo Scientific Chemicals

Tetraphenylphosphonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A11539.22
100 g, Each
Quantity
Catalog number: A11539.22
also known as A11539-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2065-67-0
IUPAC Name
tetraphenylphosphanium iodide
Molecular Formula
C24H20IP
InChI Key
AEFPPQGZJFTXDR-UHFFFAOYSA-M
SMILES
[I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or needles
Appearance (Color)
White to yellow or cream to pale brown
Assay (Titration ex Iodide)
≥98.0 to ≤102.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text