Thermo Scientific Chemicals

(+)-5-Iodo-2'-deoxyuridine, 98%

Catalog number: A11542.03
1 g, Each
Thermo Scientific Chemicals

(+)-5-Iodo-2'-deoxyuridine, 98%

Catalog number: A11542.03
1 g, Each
Quantity
Catalog number: A11542.03
also known as A11542-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54-42-2
IUPAC Name
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C9H11IN2O5
InChI Key
XQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILES
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
Appearance (Color)
White to pale cream
Assay (HPLC)
≥97.5%
Optical Rotation
30.5 ± 1.5? (C=1 in 1M NaOH)
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text