Thermo Scientific Chemicals

Quinoline, 96%, Thermo Scientific Chemicals

Catalog number: A11545.36
500 g, Each
Thermo Scientific Chemicals

Quinoline, 96%, Thermo Scientific Chemicals

Catalog number: A11545.36
500 g, Each
Quantity
Catalog number: A11545.36
also known as A11545-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-22-5
IUPAC Name
quinoline
Molecular Formula
C9H7N
InChI Key
SMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILES
C1=CC=C2N=CC=CC2=C1
Appearance (Color)
Clear colorless to pale yellow to yellow to brown to dark brown
Form
Liquid
Assay (GC)
≥95.0%
Comment
Material sourced in the U.S. and in other countries
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

Quinoline is used as a chemical intermediate for making medicines and other organic compounds.  It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline is used as a chemical intermediate for making medicines and other organic compounds.  It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.

Solubility
More soluble in hot than cold water. Miscible with alcohol, ether

Notes
Dissolves sulfur, As{2}O{3}, phosphorus
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text