m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Xylylene dichloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11548.14
also known as A11548-14
Price (USD)/ Each
81.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
81.20
Add to cart
m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
Catalog numberA11548.14
Price (USD)/ Each
81.20
-
Add to cart
Chemical Identifiers
CAS626-16-4
IUPAC Name1,3-bis(chloromethyl)benzene
Molecular FormulaC8H8Cl2
InChI KeyGRJWOKACBGZOKT-UHFFFAOYSA-N
SMILESClCC1=CC(CCl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)32.0-38.0?C
FormCrystals or powder or crystalline powder or fused/lumpy solid
Appearance (Color)White to pale yellow
Assay (GC)≥96.0%
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Joseph J. Gajewski.; Gitendra C. Paul. Photolysis of chloride precursors as a method to prepare and characterize the triplet state of fluorenyl-substituted m-xylylene diradicals. Tetrahedron Letters. 1998, 39, (5-6), 351-354.
  2. Bradford B. Wright.; Matthew S. Platz. Electron spin resonance spectroscopy of the triplet state of m-xylylene. J. Am. Chem. Soc. 1983, 105, (3), 628-630.