Thermo Scientific Chemicals

m-Xylylene dichloride, 97%, Thermo Scientific Chemicals

Catalog number: A11548.22
100 g, Each
Thermo Scientific Chemicals

m-Xylylene dichloride, 97%, Thermo Scientific Chemicals

Catalog number: A11548.22
100 g, Each
Quantity
Catalog number: A11548.22
also known as A11548-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-16-4
IUPAC Name
1,3-bis(chloromethyl)benzene
Molecular Formula
C8H8Cl2
InChI Key
GRJWOKACBGZOKT-UHFFFAOYSA-N
SMILES
ClCC1=CC(CCl)=CC=C1
Melting Point (clear melt)
32.0-38.0?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Appearance (Color)
White to pale yellow
Assay (GC)
≥96.0%

Description

m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text