2',4',6'-Trimethylacetophenone, 97+%, Thermo Scientific Chemicals
2',4',6'-Trimethylacetophenone, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',4',6'-Trimethylacetophenone, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11549.14
also known as A11549-14
Price (USD)/ Each
98.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
98.70
Add to cart
2',4',6'-Trimethylacetophenone, 97+%, Thermo Scientific Chemicals
Catalog numberA11549.14
Price (USD)/ Each
98.70
-
Add to cart
Chemical Identifiers
CAS1667-01-2
IUPAC Name1-(2,4,6-trimethylphenyl)ethan-1-one
Molecular FormulaC11H14O
InChI KeyXWCIICLTKWRWCI-UHFFFAOYSA-N
SMILESCC(=O)C1=C(C)C=C(C)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5150-1.5190 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>97.0%
2',4',6'-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Eva Hevia.; Kenneth W. Henderson.; Alan R. Kennedy.; Robert E. Mulvey. Synthesis and Characterization of New Mixed-Metal Sodium-Magnesium Enolates Derived from 2,4,6-Trimethylacetophenone. Organometallics. 2006, 25, (7), 1778-1785.
  2. Kresge, A.J. Meng, Q. 2,4,6-Trimethylthioacetophenone and Its Enol. The First Quantitative Characterization of a Simple Thiocarbonyl System in Aqueous Solution. J. Am. Chem. Soc. 1998, 120, (45), 11830-11831.