Thermo Scientific Chemicals

ACES, 99%

Catalog number: A11553.14
25 g, Each
Thermo Scientific Chemicals

ACES, 99%

Catalog number: A11553.14
25 g, Each
Quantity
Catalog number: A11553.14
also known as A11553-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7365-82-4
IUPAC Name
2-[(carbamoylmethyl)amino]ethane-1-sulfonic acid
Molecular Formula
C4H10N2O4S
InChI Key
DBXNUXBLKRLWFA-UHFFFAOYSA-N
SMILES
NC(=O)CNCCS(O)(=O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

ACES is used as a Good's buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ACES is used as a Good′s buffer is an ideal separator in the resolution of protein systems, a zwitterionic compound used in biochemical and biological research.

Solubility
Soluble in water(18g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text