Thermo Scientific Chemicals

Dibenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11554.0B
1000 g, Each
Thermo Scientific Chemicals

Dibenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11554.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11554.0B
also known as A11554-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-49-1
IUPAC Name
dibenzylamine
Molecular Formula
C14H15N
InChI Key
BWLUMTFWVZZZND-UHFFFAOYSA-N
SMILES
C(NCC1=CC=CC=C1)C1=CC=CC=C1
Refractive Index
1.5725-1.5765 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%

Description

Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

Solubility
Soluble in water(0.4g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text