Thermo Scientific Chemicals

3,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11558.22
100 g, Each
Thermo Scientific Chemicals

3,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11558.22
100 g, Each
Quantity
Catalog number: A11558.22
also known as A11558-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139-85-5
IUPAC Name
3,4-dihydroxybenzaldehyde
Molecular Formula
C7H6O3
InChI Key
IBGBGRVKPALMCQ-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=O)C=C1O
Form
Powder
Appearance (Color)
Pale brown to pale gray
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
151.0-158.0?C
Identification (FTIR)
Conforms

Description

3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

Solubility
Soluble in water(6.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text