Thermo Scientific Chemicals

Diphenylphosphinic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11563.06
5 g, Each
Thermo Scientific Chemicals

Diphenylphosphinic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11563.06
5 g, Each
Quantity
Catalog number: A11563.06
also known as A11563-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1707-03-5
IUPAC Name
diphenylphosphinic acid
Molecular Formula
C12H11O2P
InChI Key
BEQVQKJCLJBTKZ-UHFFFAOYSA-N
SMILES
OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Melting Point (clear melt)
191.0-198.5°C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5% (non-U.S. sourced material)

Description

Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text