Thermo Scientific Chemicals

Diethyl 2-(2-cyanoethyl)malonate, 98+%, Thermo Scientific Chemicals

Catalog number: A11567.18
50 g, Each
Thermo Scientific Chemicals

Diethyl 2-(2-cyanoethyl)malonate, 98+%, Thermo Scientific Chemicals

Catalog number: A11567.18
50 g, Each
Quantity
Catalog number: A11567.18
also known as A11567-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17216-62-5
IUPAC Name
1,3-diethyl 2-(2-cyanoethyl)propanedioate
Molecular Formula
C10H15NO4
InChI Key
YJJLOESDBPRZIP-UHFFFAOYSA-N
SMILES
CCOC(=O)C(CCC#N)C(=O)OCC
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4345-1.4385 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text