Thermo Scientific Chemicals

Oxamide, 98+%, Thermo Scientific Chemicals

Catalog number: A11570.22
100 g, Each
Thermo Scientific Chemicals

Oxamide, 98+%, Thermo Scientific Chemicals

Catalog number: A11570.22
100 g, Each
Quantity
Catalog number: A11570.22
also known as A11570-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-46-5
IUPAC Name
ethanediamide
Molecular Formula
C2H4N2O2
InChI Key
YIKSCQDJHCMVMK-UHFFFAOYSA-N
SMILES
NC(=O)C(N)=O
Appearance (Color)
White
Elemental Analysis
Carbon C: 26.73 - 27.82% (Theory 27.28%)
Form
powder
Assay (HPLC)
> 98.0%
Elemental Analysis
Nitrogen N: 31.17 - 32.45% (Theory 31.81%)

Description

Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text