Thermo Scientific Chemicals

4-Chlorobenzamidine hydriodide, 96%, Thermo Scientific Chemicals

Catalog number: A11571.03
1 g, Each
Thermo Scientific Chemicals

4-Chlorobenzamidine hydriodide, 96%, Thermo Scientific Chemicals

Catalog number: A11571.03
1 g, Each
Quantity
Catalog number: A11571.03
also known as A11571-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115297-57-9
IUPAC Name
[amino(4-chlorophenyl)methylidene]azanium
Molecular Formula
C7H8ClN2
InChI Key
LFCUMSZYMJRUHD-UHFFFAOYSA-O
SMILES
NC(=[NH2+])C1=CC=C(Cl)C=C1
Appearance (Color)
Pale yellow to yellow to pale brown
Form
Powder
Assay (Titration ex Iodide)
≥95.0 to ≤105.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text