Thermo Scientific Chemicals

3-Bromopropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11573.36
500 g, Each
Thermo Scientific Chemicals

3-Bromopropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11573.36
500 g, Each
Quantity
Catalog number: A11573.36
also known as A11573-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-92-1
IUPAC Name
3-bromopropanoic acid
Molecular Formula
C3H5BrO2
InChI Key
DHXNZYCXMFBMHE-UHFFFAOYSA-N
SMILES
OC(=O)CCBr
Appearance (Color)
Pale cream to cream to yellow to orange to brown
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
55.0-64.0?C
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

Solubility
Soluble in water, alcohol, ether, chloroform, benzene

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text