Thermo Scientific Chemicals

Sodium dodecatungstophosphate hydrate, Thermo Scientific Chemicals

Catalog number: A11575.14
25 g, Each
Thermo Scientific Chemicals

Sodium dodecatungstophosphate hydrate, Thermo Scientific Chemicals

Catalog number: A11575.14
25 g, Each
Quantity
Catalog number: A11575.14
also known as A11575-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51312-42-6
IUPAC Name
Sodium dodecamolybdophosphate hydrate
Molecular Formula
Na3O40PW12
SMILES
*
Molecular Weight (g/mol)
2946.12
Form
Crystals or powder or crystalline powder
Elemental Analysis
Tungsten (W) ≥ 68.5%
Appearance (Color)
White
Elemental Analysis
Sodium ≥ 1.25%
Solution Test
100mg/ml water: clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text