Thermo Scientific Chemicals

Bromocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: A11593.0E
2500 g, Each
Thermo Scientific Chemicals

Bromocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: A11593.0E
2500 g, Each
Quantity
Catalog number: A11593.0E
also known as A11593-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-85-0
IUPAC Name
bromocyclohexane
Molecular Formula
C6H11Br
InChI Key
AQNQQHJNRPDOQV-UHFFFAOYSA-N
SMILES
BrC1CCCCC1
Refractive Index
1.4940-1.4980 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & Light Sensitive. Store away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text