Thermo Scientific Chemicals

Triphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A11605.09
10 g, Each
Thermo Scientific Chemicals

Triphenylsilane, 97%, Thermo Scientific Chemicals

Catalog number: A11605.09
10 g, Each
Quantity
Catalog number: A11605.09
also known as A11605-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
789-25-3
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
42.0-48.0?C

Description

Triphenylsilane acts as a reactant or reagent for catalytic hydrogen deuterium exchange reactions of silanes, to be oxidized by carbon nanotube-gold nanohybrids, for hydrolysis by ruthenium complexes, for hydrosilylation to produce enolsilanes, for synthesis of bromosilanes, for ozone oxidation of silyl-alkenes for synthesis of α-O-silylated acyloin derivatives. It is used to synthesize hydrido silyl and bis(silyl) bis(imidazolinylidene) nickel complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsilane acts as a reactant or reagent for catalytic hydrogen deuterium exchange reactions of silanes, to be oxidized by carbon nanotube-gold nanohybrids, for hydrolysis by ruthenium complexes, for hydrosilylation to produce enolsilanes, for synthesis of bromosilanes, for ozone oxidation of silyl-alkenes for synthesis of α-O-silylated acyloin derivatives. It is used to synthesize hydrido silyl and bis(silyl) bis(imidazolinylidene) nickel complexes.

Solubility
Soluble in methanol. Decomposes in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text