2-n-Pentylfuran, 98%, Thermo Scientific Chemicals
2-n-Pentylfuran, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-n-Pentylfuran, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11607.09
also known as A11607-09
Price (USD)/ Each
48.65
Online exclusive
53.60 
Save 4.95 (9%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
53.60 
Save 4.95 (9%)
Add to cart
2-n-Pentylfuran, 98%, Thermo Scientific Chemicals
Catalog numberA11607.09
Price (USD)/ Each
48.65
Online exclusive
53.60 
Save 4.95 (9%)
-
Add to cart
Chemical Identifiers
CAS3777-69-3
IUPAC Name2-pentylfuran
Molecular FormulaC9H14O
InChI KeyYVBAUDVGOFCUSG-UHFFFAOYSA-N
SMILESCCCCCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4470-1.4510 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only