Thermo Scientific Chemicals

2,3-Dimethoxytoluene, 98+%, Thermo Scientific Chemicals

Catalog number: A11617.06
5 g, Each
Thermo Scientific Chemicals

2,3-Dimethoxytoluene, 98+%, Thermo Scientific Chemicals

Catalog number: A11617.06
5 g, Each
Quantity
Catalog number: A11617.06
also known as A11617-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4463-33-6
IUPAC Name
1,2-dimethoxy-3-methylbenzene
Molecular Formula
C9H12O2
InChI Key
WMXFNCKPYCAIQW-UHFFFAOYSA-N
SMILES
COC1=CC=CC(C)=C1OC
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.5125-1.5165 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

Solubility
Soluble in Dichloromethane. Not miscible in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text